Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ PBS Tablets
SDP

Catalog No. NB90062086 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 Tablets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB90062086 100 Tablets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB90062086 Supplier R&D Systems™ Supplier No. NB90062086

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Phosphate buffered saline tablets pH 7.4

Dilutions

  • Western Blot
  • Immunocytochemistry/ Immunofluorescence
  • Immunohistochemistry 1:10-1:500
  • Immunohistochemistry- Frozen
  • Immunohistochemistry- Paraffin

Applications: Western Blot, Immunocytochemistry/ Immunofluorescence

Recommended Storage

Store at room temperature

Specifications

Chemical Name or Material Phosphate Buffered Saline (PBS)
Product Line Novus
Quantity 100 Tablets
Grade Not Available
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.