Learn More
Aviva Systems Biology Corp PBX3 Antibody - N-terminal region (ARP32070_T100) | 100uL
Supplier: Aviva Systems Biology Corp ARP32070_T100
This is a rabbit polyclonal antibody against PBX3. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human PBX3. Alternate names: RP11-336P12.1ApplicationIHC, WB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Rat, Zebrafish
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.