Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCB Kit #1, Restek™
SDP

Catalog No. 06719737 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Concentration:
PCB Kit, Aroclors, 1000ug/mL, hexane, 1mL/ampul
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
06-719-737 PCB Kit, Aroclors, 1000ug/mL, hexane, 1mL/ampul
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-719-737 Supplier Restek Corporation Supplier No. 32089
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Aroclor standards may exhibit lot-to-lot variation.

Compounds:

  • 200μg/mL each in isooctane, 1 mL/ampul
  • 32064: Aroclor 1016
  • 32065: Aroclor 1221
  • 32066: Aroclor 1232
  • 32067: Aroclor 1242
  • 32068: Aroclor 1248
  • 32069: Aroclor 1254
  • 32070: Aroclor 1260

Specifications

CAS 12674-11-2, 110-54-3
CAS Max % 99.900000
CAS Min % 0.100000%
Odor Mild
Vapor Density 2.97 (Air = 1)
Specific Gravity 0.672 g/cm3 at 15°C
Concentration PCB Kit, Aroclors, 1000ug/mL, hexane, 1mL/ampul
UN Number UN1208
Melting Point -95°C
Solubility Negligible
Type Multi-component Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.