Learn More
MBL International Corporation Anti-PCBP1 (Human) pAb (Polyclonal Antibody)
SDP
Supplier:  MBL International Corporation RN024P
Encompass_Preferred
Clonality:Polyclonal; Application:IP|RIP|WB; Conjugate:Unlabeled; Species Reactivity:Human|Mouse|Rat; Size:200 µl; Target:PCBP1; Polyclonal Antibody of 200 targeting PCBP1 for IP, RIP, WB.PCBP1 Polyclonal Antibody.
Catalog No. 50-510-43
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.