Learn More
Aviva Systems Biology Corp PCDHA10 Antibody - N-terminal region (ARP49344_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP49344_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PCDHA10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PCDHA10. Alternate names: CNR8, CNRN8, CNRS8, CRNR8, PCDH-ALPHA10ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60069