Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ PCDHB16 (Human) Recombinant Protein (P01)

Catalog No. 89968733 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-968-733 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-968-733 Supplier Abnova™ Supplier No. H00057717P01
Add to Cart
Add to Cart

Human Recombinant Protein

Human PCDHB16 full-length ORF ( AAH36062.1, 1 a.a. - 776 a.a.) recombinant protein with GST-tag at N-terminal.

Specifications

Accession Number AAH36062.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50 mM Tris-HCI, 10 mM reduced Glutathione, pH=8.0 in. the elution buffer
Gene ID (Entrez) 57717
Name protocadherin beta 16
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue
Quantity 2 μg
Source Wheat Germ (in vitro)
Storage Requirements Store at -80°C Aliquot to avoid repeated freezing and thawing
Gene Alias KIAA1621,ME1,PCDH-BETA16,PCDH3X,PCDHB8a
Common Name PCDHB16
Recombinant Recombinant
Sequence MEIGWMHNRRQRQVLVFFVLLSLSGAGAELGSYSVVEETERGSFVANLGKDLGLGLTEMSTRKARIISQGNKQHLQLKAQTGDLLINEKLDREELCGPTEPCILHFQVLMENPLEIFQAELRVIDINDHSPMFTEKEMILKIPENSPLGTEFPLNHALDLDVGSNNVQNYKISPSSHFRVLIHEFRDGRKYPELVLDKELDREEEPQLRLTLTALDGGSPPRSGTAQVRIEVVDINDNAPEFEQPIYKVQIPENSPLGSLVATVSARDLDGGANGKISYTLFQPSEDISKTLEVNPMTGEVRLRKQVDFEMVTSYEVRIKATDGGGLSGKCTLLLQVVDVNDNPPQVTMSALTSPIAENSPEIVVAVFSVSDPDSGNNGKTISSIQEDLPFLLKPSVKNFYTLVTERALDREARAEYNITLTVTDMGTPRLKTEHNITVQISDVNDNAPTFTQTSYTLFVRENNSPALHIGSVSATDRDSGTNAQVTYSLLPPQDPHLPLASLVSINADNGHLFALRSLDYEALQAFEFRVGATDRGSPALSREALVRVLVLDANDNSPLVLYPLQNGSAPCTELVPRAAEPGYLVTKVVAVDGDSGQNAWLSYQLLKATEPGLFGVWAHNGEVRTARLLSERDAAKQRLVVLVKDNGEPPRSATATLHVLLVDGFSQPFLPLPEAAPGQTQANSLTVYLVVALASVSSLFLFSVLLFVAVRLCRRSRAASVGRCSMPEGPFPGRLVDVSGTGTLSQSYQYEVCLTGGSETSEFKFLKPIIPNFSP
Target Species Named Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.