Learn More
ATCC pcDNA3L-His-CAMYEL plasmid in Escherichia coli - Mammalian expression vector encoding CAMYEL BRET cAMP sensor
Supplier: ATCC MBA277
pcDNA3L-His-CAMYEL plasmid is a mammalian expression vector encoding the CAMYEL cAMP sensor (YFP-Epac-RLuc) used for BRET-based monitoring of intracellular cAMP dynamics in cell signaling studies. The plasmid is maintained in Escherichia coli and provided for molecular biology workflows involving BRET assays.
- Encodes CAMYEL BRET cAMP sensor for real-time cAMP monitoring.
- Designed for expression in mammalian cells to support signaling studies.
- Maintained in Escherichia coli Stbl2 for plasmid propagation.
- Contains neomycin and ampicillin selection markers for selection in eukaryotic and bacterial systems.
- Construct size approximately 9.38 kb.
- Sequence data is provided to support experimental design and cloning.
- Requires long-term storage at -80°C to preserve construct integrity.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.