Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102827
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102827 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102827 Supplier Invitrogen™ Supplier No. PA5102827
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PCM1.

PCM-1 is required for centrosome assembly and function. This protein is essential for the correct localization of several centrosomal proteins including CEP250, CETN3, PCNT and NEK2. The protein is required to anchor microtubules to the centrosome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCM1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PCM1
Gene Accession No. Q15154, Q9R0L6
Gene Alias 2600002H09Rik; 9430077F19Rik; C030044G17Rik; hPCM-1; mPCM-1; Pcm1; PCM-1; pericentriolar material 1; pericentriolar material 1 protein; pericentriolar material 1, PCM1; pericentriolar material gene 1; PTC4; RET/PCM-1
Gene Symbols PCM1
Host Species Rabbit
Immunogen A synthesized peptide derived from human PCM1(Accession Q15154), corresponding to amino acid residues N108-T158.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18536, 5108, 81740
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.