Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCOTH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA524138
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA524138 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA524138 Supplier Invitrogen™ Supplier No. PA524138
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PCOTH Polyclonal Antibody for Western Blot, IHC (P), Flow

May be involved in growth and survival of prostate cancer cells through the TAF-Ibeta pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCOTH
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene PCOTH
Gene Accession No. Q58A44
Gene Alias C1QTNF9B antisense gene protein 1; C1QTNF9B antisense RNA 1; C1QTNF9B antisense RNA 1 (non-protein coding); C1QTNF9B-AS1; PCOTH; prostate collagen triple helix protein; protein PCOTH
Gene Symbols PCOTH
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 14-42 amino acids from the central region of human PCOTH.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 542767
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.