Learn More
Sartorius PCR Hood no air flow
Catalog No. 502891631 Shop All Sartorius Products
Encompass
If sterile working conditions are not required the CellCelector PCR hood is the perfect cost-effective solution. It protects the CellCelector from dust and debris keeping it clean for daily use. Specially designed in-built safety features of the PCR hood include access doors equipped with interlock sensors which are required for the safe operation of the instrument.
Catalog No. 50-289-1631 Supplier Sartorius Supplier No. CC6002
To request a quote for this item, please select Request a Quote to reach our dedicated support team.
If sterile working conditions are not required the CellCelector PCR hood is the perfect cost-effective solution. It protects the CellCelector from dust and debris keeping it clean for daily use. Specially designed in-built safety features of the PCR hood include access doors equipped with interlock sensors which are required for the safe operation of the instrument.