Learn More
Promega PCR Optimization Kit, 1 each
Catalog No. PRD2381 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 Each
Catalog No. Quantity
PRD2381 1 Each
1 options
This item is not returnable. View return policy
Catalog No. PRD2381 Supplier Promega Supplier No. D2381
This item is not returnable. View return policy
PCR Optimization Kit is a portfolio of preformulated high-quality buffers, 25mM MgCl2 and GoTaq MDx Hot Start Polymerase. Perform a series of short experiments to quickly survey which combination will yield optimal PCR performance specific for your assay.
  • Preformulated PCR Master Mix Components to Determine Optimal PCR Amplification Conditions
  • Use this kit to build your custom, Made−to−Order 2X PCR Master Mix
  • Manufactured under cGMP for consistent, lot−to−lot PCR performance
  • Preformulated buffers cover a wide variety of PCR performance capabilities
Specifications
Content And Storage -30°C to -10°C
Quantity 1 Each