Learn More
Mascon Medical PCR Plate, 96-well, 0.2 mL, Semi skirt, Clear, Lettering
SDP
Supplier:  Mascon Medical MM802016
Encompass_Preferred
96-Well,flat top PCR microplate is designed to fitstandard 96 well blocks.Ultra-thin and consistentwall thickness allows precise thermal transfer foroptimum results.Can be sealed with 8/12 strip capsor sealing film.96-Well PCR platesFeaturesSuitable for 0.1 mL/0.2 mL thermal cycler blocksApplication:PCR,qPCR/Real-Time PCR
Catalog No. NC3527249