Learn More
Mascon Medical Mascon Medical™ PCR Plate, 384-Well, 40μl, White, Full Skirt, 15EA/Bag, 20 Bags/Case
SDP
Supplier:  Mascon Medical FSPCR40W
Encompass_Preferred
Our Mascon Medical™ PCR Plate series is made of high-quality, medical grade polypropylene, with strong adaptability, good sealing and includes 96 and 384 wells as well as full skirt, half skirt and no skirt. The unique thin wall design improves experimental efficiency and is widely used in genetics, biochemistry, immunity, medicine and other fields. Features:•Meets ISO13485 quality management system standard•RNase/DNase free•Non-pyrogenic•Endotoxin-free•Full skirtSpecifications:•Volume: 384-well, 40μl•Qty. / Case: 300•Material: Polypropylene•Sterile: No•Size: 127.3*85*9.4mm•Color: White•Temperature Range: -80℃~120℃•Packaging Format: 15 EA/Bag, 20 Bags/Case
Catalog No. NC2458336
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.