Learn More
Boca Scientific Inc GRiSP Exo/SAP Go PCR Purification Kit, 500 reactions
Supplier:  Boca Scientific Inc GK18.0500
Encompass_Preferred
A high performance enzymatic PCR purification kit with a 15 minute protocol! This kit is fast, easy to use, and offers 100% Purification. The Exo/SAP Go PCR Purification kit is comprised of Exonuclease I (Exo I) and recombinant Shrimp Alkaline Phosphatase (rSAP) in an optimal molar ratio. Unused primers are hydrolyzed by Exo I, whilst rSAP dephosphorylates excess dNTPs. Exo/SAP Go is designed to degrade dNTPs and primers in a single reaction tube in only 5 minutes. Both enzymes are subsequently removed by heat-inactivation and DNA is ready for sequencing in just 15 minutes. No further treatment is required and recovery is 100%, even for very short PCR products.
Catalog No. 50-168-5624
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.