Learn More
Mascon Medical PCR Single Tube, 0.2 mL, Attached Flat Cap, Conical, Clear, Non-sterile, 1000 pcs/pack, 10 packs/case
SDP
Supplier:  Mascon Medical MM802001
Encompass_Preferred
PCR Tubes with Attached Flat Cap.Made of virgin polypropylene and designedfor precise fit inheat blocks to optimize heat transfer.The tubesare cleanroomproduced,ensuring that they are free from contaminants thatcould affect experimentalresults.
Catalog No. NC3527240