Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ PCR Tubes and Domed Caps, Strips of 8

Catalog No. 14230255 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
250/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-230-255 250/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-230-255

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Optimize PCR and qPCR with these 0.2 mL strip tubes, available as 8 tubes per strip in several colors or 12 tubes per strip.

  • 0.2 mL Strip Tubes are compatible with standard 0.2 mL thermal cycler blocks.
  • They are designed for both PCR and qPCR applications.
  • The caps form a secure seal, yet are easy to apply and remove.
TRUSTED_SUSTAINABILITY

Specifications

Closure Color Natural
Color Natural
Volume (Metric) Working 0.2 mL
Volume (Metric) 0.3 mL
Quantity 250/Cs.
Closure Material Polypropylene
Shape Yes
Sterility Non-sterile
Material Polypropylene
Dnase Free Yes
Closure Type Domed Strip Cap
No. of Wells 8
Closure Included Yes
Format Strip Tubes and Caps
Reaction Volume 0.2 mL
Product Type PCR Tubes
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.