Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCSK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA1057
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA1057 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA1057 Supplier Invitrogen™ Supplier No. PA1057
Only null left
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

PA1-057 detects prohormone convertase 1 (PC1) from mouse pituitary extract. PA1-057 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~80 kDa protein representing PC1 from mouse pituitary extract. The PA1-057 immunogen is a synthetic peptide corresponding to residues S(111) V Q K D S A L D L F N D P M W N(127) of mouse PC1. This sequence is 88% and 82% conserved bovine and rat, respectively. The PA1-057 immunizing peptide (Cat. # PEP-164) is available for use in neutralization and control experiments.

The protein encoded by this gene belongs to the subtilisin-like proprotein convertase family. The members of this family are proprotein convertases that process latent precursor proteins into their biologically active products. This encoded protein is a type I proinsulin-processing enzyme that plays a key role in regulating insulin biosynthesis. It is also known to cleave proopiomelanocortin, prorenin, proenkephalin, prodynorphin, prosomatostatin and progastrin. Mutations in this gene are thought to cause obesity. This encoded protein is associated with carcinoid tumors. The use of alternate polyadenylation sites has been found for this gene. Multiple alternatively spliced transcript variants have been described for this gene but their full length nature is not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCSK1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1 mg/mL BSA and 0.05% sodium azide
Gene PCSK1
Gene Accession No. P63239
Gene Alias Att-1; Bdp; BMIQ12; EC 3.4.21.93; Furin homolog; LOW QUALITY PROTEIN: neuroendocrine convertase 1; NEC 1; NEC1; Nec-1; Neuroendocrine convertase 1; PC1; PC3; PCSK1; Phpp-1; Prohormone convertase 1; prohormone convertase 1/3; prohormone convertase 3; propeptide-processing protease; Proprotein convertase 1; proprotein convertase subtilisin/kexin type 1; Protein convertase subtilisin / kexin type I; Protein convertase subtilisin / kexin, type I; SPC3
Gene Symbols PCSK1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues S(111) V Q K D S A L D L F N D P M W N(127) of mouse PC1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18548
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.