Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCSK4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552168
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552168 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552168 Supplier Invitrogen™ Supplier No. PA552168
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ENKGYYFNTG TLYRYTLLLY GTAEDMTARP TGPQVTSSAC VQRDTEGLCQ ACDGPAYILG QLCLAYCPPR FFNHTRLVTA GPGHTAAPAL RVCSSCHASC YTCRGGSPRD CTSCPPSSTL DQQQGSCMGP T Highest antigen sequence identity to the following orthologs: Mouse - 43%, Rat - 35%.

Involved in the processing of hormone and other protein precursors at sites comprised of pairs of basic amino acid residues. Plays a role in transcriptional coactivation. May be involved in stabilizing the multiprotein transcription complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCSK4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PCSK4
Gene Accession No. Q6UW60
Gene Alias AI647044; KEX2-like endoprotease 3; L21221; NEC 3; NEC3; Nec-3; Neuroendocrine convertase 3; PC4; Pcsk4; prohormone convertase 3; Proprotein convertase 4; Proprotein convertase subtilisin/kexin type 4; SPC5; testicular tissue protein Li 135; UNQ2757/PRO6496
Gene Symbols PCSK4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PCSK4. Recombinant protein control fragment (Product #RP-88823).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54760
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.