Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCSK4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579788
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579788 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579788 Supplier Invitrogen™ Supplier No. PA579788
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat brain.

Involved in the processing of hormone and other protein precursors at sites comprised of pairs of basic amino acid residues. Plays a role in transcriptional coactivation. May be involved in stabilizing the multiprotein transcription complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCSK4
Applications Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene PCSK4
Gene Accession No. P29121, Q78EH2
Gene Alias AI647044; KEX2-like endoprotease 3; L21221; NEC 3; NEC3; Nec-3; Neuroendocrine convertase 3; PC4; Pcsk4; prohormone convertase 3; Proprotein convertase 4; Proprotein convertase subtilisin/kexin type 4; SPC5; testicular tissue protein Li 135; UNQ2757/PRO6496
Gene Symbols PCSK4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the N-terminus of mouse Proprotein convertase PC4 (117-133aa DPWFSKQWYMNKEIQQD).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171085, 18551
Target Species Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.