Learn More
MBL International Corporation CircuLex PCSK9-LDLR in vitro Binding Assay Kit
SDP
Supplier:  MBL International Corporation CY8150
Encompass_Preferred
Size:96 Assays; Target:PCSK9-LDLR; The CycLex Research Product CircuLex PCSK9-LDLR in vitro Binding Assay Kit can be used for screening inhibitors of PCSK9-LDLR interaction in vitro.CircuLex PCSK9-LDLR in vitro Binding Assay Kit.
Catalog No. 50-456-21
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.