Learn More
ACROBiosystems Recombinant Protein;Human PCSK9 Protein, Fc Tag;HEK293;50UG;PC9,PCSK9,PCSK-9,FH3,HCHOLA3,LDLCQ1,NARC-1,NARC1,PC9
Supplier:  ACROBiosystems PC9H525650UG
Encompass_Preferred
Recombinant Protein;Human PCSK9 Protein, Fc Tag;50UG;Human PCSK9, Fc Tag (PC9-H5256) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gln 692 (Accession # Q8NBP7-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 97.5 kDa. The protein migrates as 100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human VLDL R, His Tag (Cat. No. VLR-H5227) at 5 μg/mL (100 μL/well) can bind Human PCSK9, Fc Tag (Cat. No. PC9-H5256) with a linear range of 1-10 ng/mL (QC tested).;Immobilized Human PCSK9, Fc Tag (Cat. No. PC9-H5256) at 5 μg/mL (100 μL/well) can bind Biotinylated Human LDL R, His Tag, Avi Tag (Cat. No. LDR-H82E7) with a linear range of 0.04-0.625 μg/mL (Routinely tested).
Catalog No. 50-165-7537
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.