Learn More
Description
PA1-545 detects ethanolaminephosphate cytidyltransferase (ECT) protein in human, rat, mouse and non-human primate samples. PA1-545 has successfully been used in Western blot procedures. By Western blot, this antibody detects an ~50 kDa protein representing ECT from C3H/10T1/2 cell lysates. PA1-545 immunizing peptide corresponds to amino acid residues 157-172 from human ECT protein. This immunizing peptide (PEP-241) can be used for neutralization and control experiments.
Specifications
Specifications
| Antigen | PCYT2 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | tris with 1mg/mL BSA, glycine and 0.02% thimerosal |
| Gene | PCYT2 |
| Gene Accession No. | O88637, Q922E4, Q99447 |
| Gene Alias | 1110033E03Rik; CTP:phopshoethanolamine cytidylyltransferase; CTP:phosphoethanolamine cytidylyltransferase; ET; ethanolamine-phosphate cytidylyltransferase; PCYT2; phosphate cytidylyltransferase 2, ethanolamine; phosphorylethanolamine transferase |
| Gene Symbols | PCYT2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
