Learn More
Promega PD-1/PD-L1 Blockade Bioassay, 5X, 1 each
Catalog No. PRJ1255 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 Each
Catalog No. Quantity
PRJ1255 1 Each
1 options
Catalog No. PRJ1255 Supplier Promega Supplier No. J1255
Add to Cart
Add to Cart
Request a Quote
The PD-1/PD-L1 Blockade Bioassay is a biologically relevant MOA-based assay that can be used to measure the potency and stability of antibodies and other biologics designed to block the PD-1/PD-L1 interaction.
  • Immune Checkpoint Bioassays for Characterizing Immune−Modulating Antibodies
  • Prequalified according to ICH guidelines
  • 96− and 384−well plate formats
  • No primary cell culture required
Specifications
Content And Storage Components at different temps
Quantity 1 Each