Learn More
Selleck Chemical LLC PD-1/PD-L1 Inhibitor 3 1mg 1629654-95-0
SDP
Supplier:  Selleck Chemical LLC S81581MG
Encompass_Preferred
PD-1/PD-L1 Inhibitor 3(Programmed Death-1/Programmed Death-Ligand 1 Inhibitor 3)is a Macrocyclic inhibitor of PD-1/PD-L1 interaction with IC50 of 5.6 nM *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5727
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.