Learn More
Human PD-1 Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | PD-1 |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Clone | 2706A |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry 3-25 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. |
| Gene Alias | CD279, CD279 antigen, hPD-1, PD1, PD1hPD-l, PDCD1, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2 |
| Host Species | Rabbit |
| Immunogen | Human PD-1 synthetic peptide |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.