Learn More
AdipoGen PD 150,606 (5 mg)
SDP
Supplier:  AdipoGen AGCR10066M005
Encompass_Preferred
Chemical. CAS: 426821-41-2. Formula: C9H7IO2S. MW: 306.1. Cell permeable and selective calpain-1 and -2 inhibitor. Interacts with calcium-binding domain of calpain. Neuroprotective. Ca2+-permeable AMPA receptor inhibitor. Apoptosis inhibitor Autophagy activator by preventing cleavage of ATG5
Catalog No. 50-205-3820
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.