Learn More
Apexbio Technology LLC SC75741 913822-46-5 10mg
SDP
Supplier:  Apexbio Technology LLC B474910
Encompass_Preferred
SC75741 is a small-molecule inhibitor targeting the NF- B signaling pathway with an EC50 of approximately 200 nM NF- B is a transcription factor complex regulating immune responses through control of cytokines and chemokines production SC75741 inhibits influenza virus replication by suppressing NF- B-mediated host-dependent viral growth Experiments in MDCK cells have shown optimal suppression of virus replication at concentrations around 5 M post-infection Additionally experiments conducted in human alveolar epithelial A549 cells infected with influenza strains including H5N1 and H7N7 indicated dose-dependent reduction in viral titers upon treatment SC75741 administration in influenza-infected mice showed reduced viral load and lowered expression of cytokines such as IL-6 and IP-10 indicating its suitability for studying virus-induced inflammatory responses and replication
Catalog No. 50-000-62744
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.