Learn More
AdipoGen PD 98,059 (5 mg)
SDP
Supplier:  AdipoGen AGCR10118M005
Encompass_Preferred
Chemical. CAS: 167869-21-8. Formula: C16H13NO3. MW: 267.3. Highly selective, reversible and cell permeable MEK (MAP kinase kinase) inhibitor. Blocks the phosphorylation and activation of the MAP kinase pathway. T cell activation inhibitor. Inhibits cell growth and cell proliferation of several cancer cells.
Catalog No. 50-205-3846
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.