Learn More
CPC Scientific cyclo(AcTyr-MeAla-Asn-Pro-His-Leu-Hyp-Trp-Ser-Trp(Me)-MeNle-MeNle-Orn-Cys)-Gly-NH2 (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific CANC022A
Encompass
WL12 peptide selectively binds to programmed death ligand 1 (PD-L1), an immune checkpoint protein that is overexpressed in many cancer cell lines. Tumor PD-L1 expression contributes to immune system depression and is indicative of a tumor?s response to PD-L1 and PD-1.SEQUENCE: cyclo(AcTyr-MeAla-Asn-Pro-His-Leu-Hyp-Trp-Ser-Trp(Me)-MeNle-MeNle-Orn-Cys)-Gly-NH2 (trifluoroacetate salt)(Thioether bridge: Cys14-Ac-Tyr)ONE-LETTER SEQUENCE: cyclo(AcY-MeANPHL-Hyp-WSW(Me)-MeNle-MeNle-OC)-G-NH2 (trifluoroacetate salt)(Thioether bridge: C14-Ac-Y)MOLECULAR FORMULA: C91H128N22O20SMOLECULAR WEIGHT: 1882.19STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: CancerCITATIONS:Lesniak, Wojciech G. et al. Molecular Imaging 18 (2019)?Chatterjee
Catalog No. 50-210-8899
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.