Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PD-L1, Mouse anti-Human, Clone: 5H1, Millipore Sigma™

Catalog No. p-7162894 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC111525 25 μL
MABC1115100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABC111525 Supplier MilliporeSigma Supplier No. MABC111525UL
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-PD-L1, clone 5H1, Cat. No. MABC1115, is a mouse monoclonal antibody that detects PD-L1 and has been tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.

Programmed cell death 1 ligand 1 (UniProt Q9NZQ7; also known as B7-H1, B7 homolog 1, CD274, PD-L1, PDCD1 ligand 1, Programmed death ligand 1) is encoded by the CD274 (also known as B7H1, PDCD1L1, PDCD1LG1, PDL1) gene (Gene ID 29126) in human. PD-L1 is a single-pass type I membrane protein that plays a critical role in induction and maintenance of immune tolerance to self. It is expressed on activated T- and B-cells, dendritic cells, keratinocytes, and monocytes. Higher expression has also been reported in heart, skeletal muscle, placenta, and lung. PD-1 and PD-1 ligands 1 and 2 (PD-L1 and PD-L2) are B7:CD28 family members that regulate T cell activation and peripheral tolerance. When engaged together with the TCR, the interaction of PD-1 with its ligands delivers an inhibitory signal to T cell proliferation and cytokine production. While PD-L1 is broadly expressed in hematopoietic and nonhematopoietic cells, PD-L2 expression is highly restricted to antigen presenting cells (APCs), including dendritic cells (DCs) and macrophages. The PD-1 pathway plays a key role in the progressive loss of effector T cell responses during chronic HIV infection. Under some conditions, blockade of this pathway can restore many T cell functions. PD-L1 is initially produced with signal peptide (aa 1-18) sequence, which is subsequently cleaved off to produce the mature protein with a large extracellular (aa 19-238) region that contains an Ig-like V-type domain (aa 19-127) and an Ig-like C2-type domain (aa 133-225), followed by a transmembrane domain (aa 239-259) and a cytoplasmic tail (aa 260-290).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PD-L1
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 5H1
Conjugate Unconjugated
Dilution Immunohistochemistry (Paraffin) Analysis: A representative lot detected PD-L1 in Immunohistochemistry applications (Bigelow, E., et. al. (2013). J Vis Exp. 3;(71); Spranger, S., et. al. (2013). Sci Transl Med. 5(200):200ra116; Parra, E.R., et. al. (2018). Appl Immunohistochem Mol Morphol. 26(2):83-93; Andorsky, D.J., et. al. (2011). Clin Cancer Res. 17(13):4232-44; Sunshine, J.C., et. al. (2017). Clin Cancer Res. 23(16):4938-4944).Western Blotting Analysis: A representative lot detected PD-L1 in Western Blotting applications (Parra, E.R., et. al. (2018). Appl Immunohistochem. Mol. Morphol. 26(2):83-93_.
Formulation Purified mouse monoclonal antibody IgG1 in PBS without azide.
Gene Alias Programmed cell death 1 ligand 1;B7-H1;B7 homolog 1;CD274;PDCD1 ligand 1;Programmed death ligand 1
Gene Symbols CD274;B7H1;PDCD1L1;PDCD1LG1;PDL1
Host Species Mouse
Immunogen Full-length human recombinant PD-L1.
Purification Method Protein G purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Apoptosis & Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_054862.1
Test Specificity Clone 5H1 detects human Programmed cell death ligand 1 (PD-L1). It targets an epitope with in the extracellular domain.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.