Learn More
Sino Biological Recombinant Human PD-L1/B7-H1 Protein (ECD, hFc Tag), HPLC-verified, Amine-Labeled Biotinylated
Supplier:  Sino Biological 10084H02HB100
Encompass_Preferred
A DNA sequence encoding the human CD274 (NP_054862.1) (Met1-Thr239) was expressed with C-terminal fused Fc region of human IgG1. The purified protein was biotinylated in vitro.
Catalog No. 50-211-6193