Learn More
Sino Biological Human PD-L1 / B7-H1 / CD274 Protein (His Tag) , Biotinylated 100ug
Supplier:  Sino Biological 10084H08HB100
Encompass_Preferred
A DNA sequence encoding the N-terminal segment (Met1-Thr239) of the extracellular domain of human B7-H1 (NP_054862.1) was expressed with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.
Catalog No. 50-211-6192
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.