Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PD-L1 Recombinant Human Monoclonal Antibody (332M7)
GREENER_CHOICE

Catalog No. MA551820
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA551820 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA551820 Supplier Invitrogen™ Supplier No. MA551820
Add to Cart
Add to Cart

Human Recombinant Monoclonal Antibody

This antibody strongly binds human and cynomolgus monkey PD-L1 and shows weak binding for mouse PD-L1. PD-L1 is highly expressed in many human tumors. . PD-L1 exerts an immune suppressive signal through binding to PD-1 and B7.1, and tumor expression of PD-L1 can mediate cancer immune evasion. Blocking this pathway, therefore provides a rationale for cancer immunotherapy.

Programmed death receptor ligand 1 (PD-L1, also called B7-H1) is a recently described B7 family member. To date, one specific receptor has been identified that can be ligated by PD-L1. This receptor, programmed death receptor 1 (PD-1), has been shown to negatively regulate T-cell receptor (TCR) signaling. Upon ligating its receptor, PD-L1 has been reported to decrease TCR-mediated proliferation and cytokine production. PD-L1 expression was found to be abundant on many murine and human cancers and could be further up-regulated upon IFN-gamma stimulation. Thus, PD-L1 might play an important role in tumor immune evasion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PD-L1
Applications Flow Cytometry, Inhibition Assays
Classification Recombinant Monoclonal
Clone 332M7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene Cd274
Gene Accession No. Q9EP73, Q9NZQ7
Gene Alias A530045L16Rik; B7 homolog 1; B7-H; B7h1; B7-H1; Cd274; CD274 antigen; CD274 molecule; PDCD1 ligand 1; Pdcd1l1; Pdcd1lg1; Pdl1; PD-L1; pdl-1; Programmed cell death 1 ligand 1; programmed death ligand 1; RGD1566211; sCD274; soluble CD274; soluble PD L1; sPD L1
Gene Symbols Cd274
Host Species Human
Immunogen The original parental antibody 332M1 was generated by immunizing BALB/c mice with recombinant human PD-L1. Later on the humanized version of 332M1 was generated (332M7) by grafting the CDRs of the original mouse antibody onto human framework regions.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 102145573, 29126, 60533
Target Species Human, Mouse, Cynomolgus Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.