Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PD-L1/TGF beta Biosimilar Recombinant Human Monoclonal Antibody
GREENER_CHOICE

Catalog No. MA560149
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA560149 100 μg
MA560150 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA560149 Supplier Invitrogen Supplier No. MA560149
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PD-L1/TGF beta Biosimilar Recombinant Monoclonal Antibody

PD-L1/TGF beta Biosimilar Recombinant Monoclonal Antibody for SPR, Flow, ELISA, Functional

TRUSTED_SUSTAINABILITY

Specifications

Antigen PD-L1/TGF beta Biosimilar
Applications ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Classification Recombinant Monoclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Host Species Human
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.