Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PD-L2/B7-DC/PDCD1LG2 Antibody (988708), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB350087
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB350087 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB350087 Supplier R&D Systems Supplier No. FAB12241MFV500

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

PD-L2/B7-DC/PDCD1LG2 Monoclonal specifically detects PD-L2/B7-DC/PDCD1LG2 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen PD-L2/B7-DC/PDCD1LG2
Applications Flow Cytometry
Classification Monoclonal
Clone 988708
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias B7-DC, bA574F11.2, Btdc, Butyrophilin B7-DC, CD273 antigen, CD273PD-1 ligand 2, MGC142240, PD-1-ligand 2, PDCD1L2MGC142238, PDL2B7DC, PD-L2PDCD1 ligand 2, programmed cell death 1 ligand 2, Programmed death ligand 2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human PD-L2/B7-DC, Met1-Pro219, Accession # Q9BQ51
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 80380
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.