Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PD-L2 (PDCD1LG2) Mouse anti-Human, Clone: OTI2E7, lyophilized, TrueMAB™
SDP

Catalog No. 501669217
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-9217 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-9217 Supplier OriGene Supplier No. CF809004
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

CD273 (PDCD1 Ligand 2, Programmed cell death 2 ligand 2, PD-L2, B7-DC) is involved in cell-mediated immune responses initiated by T lymphocytes that are stimulated by cognate peptides bound to MHC molecules on antigen-presenting cells (APC). T-cell activation is generally self-limited as activated T cells express receptors such as CD273 that mediate inhibitory signals from the APC (antigen presenting cell). CD273 can bind two different but related ligands, PDL-1 and PDL-2, both of which are thought act as a negative regulator of T cell activation. However, it has been suggested that PDL-2 can act to stimulate an immunogenic response through and alternative receptor from PD-1. At least three isoforms of PDL-2 are known to exist; CD273 is specific to the largest isoform. Diseases associated with CD273 include cysticercosis, and myasthenic syndrome.

Specifications

Antigen PD-L2 (PDCD1LG2)
Applications Western Blot
Classification Monoclonal
Clone OTI2E7
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene PDCD1LG2
Gene Accession No. Q9BQ51
Gene Alias B7DC, Btdc, CD273, PD-L2, PDCD1L2, PDL2, bA574F11.2
Gene Symbols PDCD1LG2
Host Species Mouse
Immunogen Full length human recombit protein of human PDCD1LG2 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80380
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.