Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva PD MidiTrap™ G-10

Catalog No. 45002062 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-002-062 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-002-062 Supplier Cytiva Supplier No. 28918011
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cleanup biological samples, such as peptides, small proteins, and oligosaccharides. Cytiva Life Sciences™ PD MidiTrap™ G-10 offers fast desalting and efficient removal of contaminants such as salts, dyes, and radioactive labels.

  • Prepacked, single-use gravity columns for buffer exchange and cleanup of biological samples, for example peptides, small proteins, and oligosaccharides
  • Bed volume: 5.3mL
  • Bed dimensions: 13 x 40mm
  • Sample volume: 400 to 1000μL
Media:
  • Matrix: cross-linked dextran
  • Particle size wet: 55 to 165μm
  • Partical size dry: 40 to 120μm
  • pH stability working range: 2 to 13
Column:
  • Column I.D.: 13mm
  • Column material: polypropylene
  • Bed support material: polyethylene

TRUSTED_SUSTAINABILITY

Specifications

Buffer 1M Acetate, pH 6, 0.1M NaOH, 70% Ethanol or 6M Guanidine Hydrochloride
Product Type PD MidiTrap G-10 gravity column
Bed Volume 5.3mL
Diameter (English) Inner 0.51 in.
Diameter (Metric) Inner 13mm
Quantity Each
Height (English) 1.5 in.
Height (Metric) 40mm
Flow Rate 24 to 49cm/hr.
Includes Instructions
Material Polyethylene
pH Range 2 to 13
Particle Size 40 to 120μm in dry form, 55 to 165μm in wet form
Support Type Cross-linked Dextran
Content And Storage 4° to 30°C
Column Type Gravity Column
For Use With (Application) Buffer exchange and cleanup of biological samples, for example Peptides, small Proteins and Oligosaccharides
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.