Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva PD MidiTrap™ G-25 Sample Preparation Columns

Catalog No. 45001530 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-001-530 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-001-530 Supplier Cytiva Supplier No. 28918008
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Four centrifuge adapters

Use for cleanup of biomolecules with a molecular weight >5000 Mr in sample volumes up to 1mL. Cytiva Life Sciences™ PD MidiTrap™ G-25 Sample Preparation Columns are convenient and prepacked.

  • Developed for a wide range of applications: desalting, buffer exchange, removal of low-molecular weight compounds, etc.
  • Equally suited for gravity flow or centrifugation protocols
  • Accommodates sample volumes of 0.5 to 1.0mL for gravity, 0.75 to 1.0mL with centrifugation
  • High recoveries of 70 to 90%, depending on biomolecule
  • High desalting capacityof >90%

TRUSTED_SUSTAINABILITY

Specifications

Product Type PD MidiTrap Sephadex G-25 gravity column
Bed Volume 8.5mL
Diameter (English) Inner 0.51 in.
Diameter (Metric) Inner 13mm
Quantity Each
Height (English) 1.02 in.
Height (Metric) 26mm
Flow Rate >100cm/hr.
Includes Four centrifuge adapters, instructions
Material Polyethylene
pH Range 2 to 13
Particle Size 50μm in dry form, 38 to 235μm in wet form
Support Type Cross-linked Dextran
Content And Storage 4° to 30°C
Column Type Gravity Column
For Use With (Application) Convenient and rapid cleanup with high reproducibility and the potential for high throughput by running multiple samples in parallel
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.