Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva PD MiniTrap™ G-10 Sample Preparation Columns
GSA_VA

Catalog No. 45001531 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-001-531 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-001-531 Supplier Cytiva Supplier No. 28918010
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Use for buffer exchange and cleanup of biological samples, for example peptides, small proteins, and oligosaccharides. Cytiva Life Sciences™ PD MiniTrap™ G-10 Sample Preparation Columns are prepacked, single-use gravity columns.

  • Prepacked with Sephadex™ G-10 medium for efficient removal of contaminants such as salts, dyes, and radioactive labels
  • High desalting capacity; no need for a purification system
  • Preparation of samples in the volume range 100 to 300μL
  • Stable in the presence of all commonly used buffers

Sample clean up prior to liquid chromatography, electrophoresis and MS

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Buffer 0.2M NaOH, 0.2M HCl, 1M Acetic Acid, 8M Urea, 6M Guanidine HCl, 1% SDS , 24% Ethanol, 30% Porpanol, 30% Aceto nitrile
Product Type PD MiniTrap Sephadex G-10 gravity column
Bed Volume 2.1mL
Diameter (English) Inner 0.38 in.
Diameter (Metric) Inner 9.7mm
Quantity Each
Height (English) 1.2 in.
Height (Metric) 28mm
Flow Rate 24 to 49cm/hr.
Includes Instructions
Material Polyethylene
pH Range 2 to 13
Particle Size 40 to 120μm in dry form, 55 to 165μm in wet form
Support Type Cross-linked Dextran
Content And Storage 4° to 30°C
Column Type Gravity Column
For Use With (Application) Buffer exchange and cleanup of biological samples, for example Peptides, small Proteins and Oligosaccharides
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.