Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva PD MultiTrap™ Plates

Catalog No. 45001528 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
4 x 96 Well Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-001-528 4 x 96 Well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-001-528 Supplier Cytiva Supplier No. 28918006
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Convenient and fast small-scale cleanup of proteins and other large biomolecules (>5000 Mr) with high reproducibility from well to well and from plate to plate

  • Designed for high-throughput desalting and buffer exchange of multiple samples
  • Range of other applications include buffer exchange and removal of low-molecular weight compounds
  • 96-well plates prepacked with 500μL Sephadex™ G-25 Medium/well for high reproducibility
  • Suitable for automated use with automated robotic systems as well as manual use
  • Sample preparation in the range of 70 to 130μL without sample dilution
  • Stable in the pH range 2 to 13, and in the presence of all commonly used buffers

TRUSTED_SUSTAINABILITY

Specifications

Buffer 0.2M NaOH, 0.2M HCl, 1M Acetic Acid, 8M Urea, 6M Guanidine HCl, 1% SDS , 24% Ethanol, 30% Porpanol, 30% Aceto nitrile
Product Type PD Sephadex G-25 MultiTrap Plate
Bed Volume 0.07 to 0.13mL
Quantity 4 x 96 Well Plate
Height (English) 11.81 in.
Height (Metric) 300mm
Flow Rate >100cm/hr.
Material Polypropylene, Polyethylene
pH Range 2 to 13
Particle Size 50μm in dry form, 38 to 235μm in wet form
Support Type Cross-linked Dextran
Content And Storage 4° to 30°C
For Use With (Application) High-throughput desalting and Buffer exchange of multiple samples, use manually or with robotic systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.