Learn More
Selleck Chemical LLC PD318088 50mg 391210-00-7
SDP
Supplier:  Selleck Chemical LLC S156850MG
Encompass_Preferred
PD318088 is a non-ATP competitive allosteric MEK1/2 inhibitor, binds simultaneously with ATP in a region of the MEK1 active site that is adjacent to the ATP-binding site. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-868-6
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.