Learn More
PDAM, Sigma-Aldrich
Catalog No. 01258979 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
Catalog No. Quantity
01-258-979 25 mg
1 options
Catalog No. 01-258-979 Supplier MilliporeSigma Supelco Supplier No. P686325MG
Add to Cart
Add to Cart
Fluorescent diazoalkane useful as an HPLC derivatization reagent for carboxylic acid detection with better detection limit parameters and greater chemical stability than the commonly used 9-anthryldiazomethane.
For HPLC derivatization
TRUSTED_SUSTAINABILITY
Specifications
Reagent Type Ion Pairing Reagent
Quantity 25 mg