Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDCD2L Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. p-7209912 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-3081 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-3081 Supplier Proteintech Group Inc Supplier No. 207811AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Over expression of PDCD2L attenutates TNF-alpha release in Daudi cells. PDCD2L over-expression restrained proliferation of HEK293T cells. DNA/flow cytometry analysis showed that the over-expression of PDCD2L severely delays cell cycle progression at S phase. These all suggest that PDCD2L might play a role in apoptosis.

Specifications

Antigen PDCD2L
Applications Western Blot
Classification Polyclonal
Concentration 0.82 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene PDCD2L
Gene Accession No. Q8C5N5, Q9BRP1
Gene Alias PDCD2L, programmed cell death 2 like
Gene Symbols Pdcd2l
Host Species Rabbit
Immunogen PDCD2L Fusion Protein Ag14795
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 68079, 84306
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.