Learn More
Invitrogen™ PDE11A Polyclonal Antibody

Description
Recommended positive controls: Rat testis, skeletal muscle, kidney and brain.
Specifications
Specifications
| Antigen | PDE11A |
| Applications | ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.68 mg/mL |
| Conjugate | Unconjugated |
| Formulation | Tris glycine with 30% glycerol, 0.5% BSA and 0.02% Sodium azide; pH 7.4-7.9 |
| Gene | PDE11A |
| Gene Accession No. | P0C1Q2, Q8VID6, Q9HCR9 |
| Gene Alias | 6330414F14Rik; A630086N24Rik; cAMP and cGMP cyclic nucleotide phosphodiesterase 11A; cAMP and cGMP phosphodiesterase 11A; Dual 3',5'-cyclic-AMP and -GMP phosphodiesterase 11A; FLJ23693; Gm350; MGC133355; MGC133356; Pde11a; PDE11A1; phosphodiesterase 11A; PPNAD2 |
| Gene Symbols | PDE11A |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.