Learn More
PDE4A Mouse anti-Human, Clone: OTI1E3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF501152

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Cyclic nucleotides are important second messengers that regulate and mediate a number of cellular responses to extracellular signals, such as hormones, light, and neurotransmitters. Cyclic nucleotide phosphodiesterases (PDEs) regulate the cellular concentrations of cyclic nucleotides and thereby play a role in signal transduction. PDE4A is a class IV cAMP-specific PDE.Specifications
PDE4A | |
Monoclonal | |
Unconjugated | |
PDE4A | |
DPDE2, PDE4, PDE46 | |
Mouse | |
Affinity Chromatography | |
RUO | |
5141 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI1E3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P27815 | |
PDE4A | |
Full length human recombit protein of human PDE4A produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.