Learn More
Bioss PDE4D Monoclonal Antibody
Supplier: Bioss BSM61841R
PDE4D Primary Antibody Unconjugated Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.