Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDE5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579796
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579796 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579796 Supplier Invitrogen™ Supplier No. PA579796
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat lung tissue, PANC-1 whole cell. IHC: human thyroid cancer tissue.

The cyclic monophosphate nucleotides (cyclic adenosine monophosphate [cAMP] and cyclic guanosine monophosphate [cGMP] are found ubiquitously in mammalian cells and act as second messenger transducers to effect the intracellular actions of a variety of G protein coupled receptors (GPCRs) for hormones, cytokines, and neurotransmitters. Cyclic nucleotides are important intracellular second messenger which play important role in a variety of signal transduction process.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDE5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene Pde5a
Gene Accession No. O54735, O76074
Gene Alias Cgbpde; CGB-PDE; cGMP-binding cGMP specific phosphodiesterase 5A2; cGMP-binding cGMP-specific 3',5'-cyclic nucleotide phosphodiesterase; cGMP-binding cGMP-specific phosphodiesterase; cGMP-binding/cGMP-specific phosphodiesterase; cGMP-specific 3',5'-cyclic phosphodiesterase; cGMP-specific phosphodiesterase PDE5A2; cGMP-specific phosphodiesterase type 5A; CN5A; Cn5n; PDE5; PDE5A; Pde5a1; PDE5A2; phosphodiesterase 5A; phosphodiesterase 5A, cGMP-specific; phosphodiesterase isozyme 5; phosphodiesterase type 5
Gene Symbols Pde5a
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the N-terminus of human PDE5A (20-63aa QKQQQRDQDSVEAWLDDHWDFTFSYFVRKATREMVNAWFAERVH).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171115, 8654
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.