Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDE6A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA1720 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA1720 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA1720 Supplier Invitrogen™ Supplier No. PA1720
  • $646.84
    $461.65 / Each of 1

    Save $185.19

    29% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

PA1-720 detects phosphodiesterase 6 alpha (PDE6 alpha) from sheep retinal extract and bovine rod outer segment membranes as well as purified bovine PDE6. PA1-720 has been successfully used in Western blot and immunofluoresence procedures. By Western blot, this antibody detects an ~90 kDa protein representing PDE6 alpha from sheep retinal extract. A higher molecular weight band is sometimes observed using sheep retinal extracts. The PA1-720 immunizing peptide corresponds to amino acid residues 1-16 from human PDE6 alpha. This sequence is completely conserved between human, mouse and bovine and 94% conserved in canine. PA1-720 immunizing peptide (Cat. # PEP-148) is available for use in neutralization and control experiments.

This gene encodes the cyclic-GMP (cGMP)-specific phosphodiesterase 6A alpha subunit, expressed in cells of the retinal rod outer segment. The phosphodiesterase 6 holoenzyme is a heterotrimer composed of an alpha, beta, and two gamma subunits. cGMP is an important regulator of rod cell membrane current, and its dynamic concentration is established by phosphodiesterase 6A cGMP hydrolysis and guanylate cyclase cGMP synthesis. The protein is a subunit of a key phototransduction enzyme and participates in processes of transmission and amplification of the visual signal. Mutations in this gene have been identified as one cause of autosomal recessive retinitis pigmentosa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDE6A
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene PDE6A
Gene Accession No. P11541, P16499
Gene Alias cGMP phosphodiesterase alpha; cGMP phosphodiesterase alpha subunit; cGMP phosphodiesterase alpha subunit (EC 3.1.4.35); CGPR-A; cyclic GMP phosphodiesterase alpha subunit; GMP phosphodiesterase alpha subunit; GMP-PDE alpha; nmf282; PDE V-B1; PDE6A; PDEA; phosphodiesterase 6A; phosphodiesterase 6A, cGMP-specific, rod, alpha; phosphodiesterase, cyclic GMP (rod receptor), alpha polypeptide; rod cGMP-specific 3',5'-cyclic phosphodiesterase subunit alpha; rod photoreceptor cGMP phosphodiesterase alpha subunit; RP43; unnamed protein product
Gene Symbols PDE6A
Host Species Rabbit
Immunogen Synthetic Peptide: M(1) G E V T A E E V E K F L D S N(16) C.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 101122024, 281973, 5145
Target Species Human, Bovine, Sheep
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.