Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDE6D Recombinant Rabbit Monoclonal Antibody (HL1359)
GREENER_CHOICE

Catalog No. PIMA547012 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547012 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547012 Supplier Invitrogen™ Supplier No. MA547012
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

PrBP/delta, previously known as PDE6/delta, is the effector enzyme in the G protein-mediated signal transduction cascade in the visual system. There are five different subunits consisting of rod and cone specific catalytic subunits: alpha (Cone), alpha (Rod), and beta (Rod), the inhibitory subunit gamma, and subunit delta of unknown function (which likely interacts with many other proteins besides the PDE6 family). PRBP/delta has been implicated in the transport and membrane targeting of prenylated proteins (including PDE6) from their site of synthesis in the inner segment to their final destination in the outer segment of rods and cones (Norton et. al., 2005).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDE6D
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone HL1359
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene PDE6D
Gene Accession No. O43924, O55057, Q9XT54
Gene Alias AI841218; GMP-PDE delta; JBTS22; PDE6D; PDED; phosphodiesterase 6D; phosphodiesterase 6D, cGMP-specific, rod, delta; PrBP/delta; Protein p17; retinal rod rhodopsin-sensitive cGMP 3',5'-cyclic phosphodiesterase subunit delta; rod phosphodiesterase delta subunit
Gene Symbols PDE6D
Host Species Rabbit
Immunogen Full length human PDE6D Recombinant protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 101096755, 18582, 363272, 403781, 436981, 5147
Target Species Human, Mouse, Rat, Canine, Feline, Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.