Learn More
Enzo Life Sciences PDGF-AA (human), (recombinant) (100µg)
Supplier: Enzo Life Sciences ENZPRT2560100
Alternative name: Platelet-derived growth factor-AA, GDGF, ODGF. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 0.1% Trifluoroacetic Acid (TFA). Source: Produced in E. coli. Contains 126/252 amino acids. Long Term Storage: -80°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.